Drukuj
Przyłbica ochronna, Speedglas™ 9100
Dostawca: 3M
501805
111-1632EA 0 PLN
111-1632
Przyłbica ochronna, Speedglas™ 9100
Osłony twarzy
This welding shield protects eyes and face from radiation, heat and sparks while providing a precise view of the work. The auto-darkening welding lens 9100XX features advanced TST (TIG Sensor Technology) capable of detecting an arc down to an industry leading 1 amp. The head harness provides multiple adjustment combinations so that users can tailor the helmet to fit their personal preferences and comfort setting. Optimised for Stick, MIG, TIG with cut, grind, tack, hidden arc and outdoor settings.
  • Welding lens 9100XX with superior optical performance for continuous viewing comfort