Drukuj
VWR® Seradigm Płodowa surowica bydlęca (FBS), ultraniskie IgG, pochodzenie US
Dostawca: VWR Seradigm
10018-826EA 5240 PLN
10018-826 10018-828 10836-696 10836-698 10018-830
VWR® Seradigm Płodowa surowica bydlęca (FBS), ultraniskie IgG, pochodzenie US
Surowice
Ultra-low IgG foetal bovine serum (FBS) is purified of immunoglobulin (IgG) using a chromatographic process that delivers significantly reduced levels of IgG (≤5 μg/ml).
  • 100% US origin
  • Triple 0,1 μm sterile filtration
  • Preservation of native, growth-promoting factors and minimisation of growth-inhibiting factors
  • Independently tested by third party laboratories
  • Fully traceable
Ultra-low IgG FBS is ideal for cell culture and protein purification applications where naturally occurring levels of IgG are too high.
Certyfikaty: International SerumWięcej informacji o produkcie