Drukuj

Peroksodisiarczan(VI) diamonu (APS) ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Dostawca: Avantor
Niebezpieczeństwo

Synonim: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

0762.1000EA 408 PLN
0762.1000 0762.5000 0762-07
Peroksodisiarczan(VI) diamonu (APS) ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Peroksodisiarczan(VI) diamonu (APS)

Uwaga: Because of inherent instability, this reagent may be expected to decrease in strength and to increase in acidity during storage.

Formuła: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Temp. topnienia: 120
Gęstość: 1.98 g/cm³ (20 °C)
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD00003390
Numer CAS: 7727-54-0
Numer WE (dawny EINECS): 231-786-5
Jednostka: 1444
ADR: 5.1 III
Klasyfikacja Merck : 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Zobacz ceny

Wyniki testów specyfikacyjnych

Meets ACS Reagent Chemical Requirements ≥98.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
Store below 20°C. ≤0.050%
Assay ((NH₄)₂S₂O₈) ≥ 98.0 %
Insoluble Matter ≤ 0.005 %
Residue after Ignition ≤ 0.050 %
Titrable Free Acid (meq/g) ≤ 0.04
Chloride and Chlorate (as Cl) ≤ 0.001 %
Heavy Metals (as Pb) ≤ 0.005 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
Trace Impurities - Manganese (Mn) ≤ 0.500 ppm
Because of inherent instability, this reagent may be expected to
decrease in strength and to increase in acidity during storage.
After storage for some time, the reagent may fail to meet the specified
requirements for Assay and Titrable Free Acid.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR